accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaONONOOSOOO

CAS RN 885-62-1

IUPAC Name: sodium 2,4-dinitrobenzene-1-sulfonate
Synonyms: DNBSO3na
Molecular Weight (g/mol): 270.15
Molecular Formula: C6H3N2NaO7S
InChi Key: GSBYVRKLPCSLNV-UHFFFAOYSA-M
SMILES: [Na+].[O-][N+](=O)C1=CC=C(C(=C1)[N+]([O-])=O)S([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results