accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 885266-67-1

IUPAC Name: [(4-nitro-1H-indol-3-yl)sulfanyl]formonitrile
Synonyms: [(4-nitro-1H-indol-3-yl)sulfanyl]formonitrile
Molecular Weight (g/mol): 219.22
Molecular Formula: C9H5N3O2S
InChi Key: FDQWHYSXVRDVMV-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C2C(SC#N)=CNC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results