accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OSOONHOOH

CAS RN 885268-78-0

IUPAC Name: 4-{[4-(2-methylphenoxy)benzenesulfonamido]methyl}benzoic acid
Synonyms: 4-{[4-(2-methylphenoxy)benzenesulfonamido]methyl}benzoic acid
Molecular Weight (g/mol): 397.45
Molecular Formula: C21H19NO5S
InChi Key: RAJRZONWHZRZEN-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1OC1=CC=C(C=C1)S(=O)(=O)NCC1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results