accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 88574-06-5

HOONHOO

CAS RN 88574-06-5

IUPAC Name: 6-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms: 6-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol): 353.42
Molecular Formula: C21H23NO4
InChi Key: FPCPONSZWYDXRD-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results