accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 885950-21-0

IUPAC Name: 3-bromoimidazo[1,2-a]pyridine-6-carbonitrile
Synonyms: 3-bromoimidazo[1,2-a]pyridine-6-carbonitrile
Molecular Weight (g/mol): 222.05
Molecular Formula: C8H4BrN3
InChi Key: UOQHWNPVNXSDDO-UHFFFAOYSA-N
SMILES: BrC1=CN=C2C=CC(=CN12)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results