CAS RN 886365-92-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(4-Cyclohexyl-phenyl)-(4-isopropyl-phenyl)-amine, TRC
CAS: 886365-92-0 Molecular Formula: C21H27N Molecular Weight (g/mol): 293.45 IUPAC Name: 4-cyclohexyl-N-(4-propan-2-ylphenyl)aniline SMILES: CC(C)c1ccc(Nc2ccc(cc2)C3CCCCC3)cc1