accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 886498-65-3

H3COFOHOF

CAS RN 886498-65-3

IUPAC Name: 2-(2,6-difluoro-3-methoxyphenyl)acetic acid
Synonyms: (2,6-difluoro-3-methoxyphenyl)acetic acid
Molecular Weight (g/mol): 202.16
Molecular Formula: C9H8F2O3
InChi Key: KIUMRKHFRAIFFF-UHFFFAOYSA-N
SMILES: COC1=C(F)C(CC(O)=O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results