accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOFFFOOH

CAS RN 886500-45-4

IUPAC Name: 2-[2-methoxy-4-(trifluoromethoxy)phenyl]acetic acid
Synonyms: [2-methoxy-4-(trifluoromethoxy)phenyl]acetic acid
Molecular Weight (g/mol): 250.17
Molecular Formula: C10H9F3O4
InChi Key: MFXSQAUASZWYDH-UHFFFAOYSA-N
SMILES: COC1=C(CC(O)=O)C=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results