accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 886502-90-5

IUPAC Name: 2,6-dichloro-4-(trifluoromethoxy)benzoic acid
Synonyms: 2,6-dichloro-4-(trifluoromethoxy)benzoic acid
Molecular Weight (g/mol): 275
Molecular Formula: C8H3Cl2F3O3
InChi Key: HJBCPDRNEWTQCH-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C=C(OC(F)(F)F)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results