accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 886851-35-0

SNNNH2

CAS RN 886851-35-0

IUPAC Name: 1-[2-(thiomorpholin-4-yl)pyridin-4-yl]methanamine
Synonyms: 1-[2-(thiomorpholin-4-yl)pyridin-4-yl]methanamine
Molecular Weight (g/mol): 209.31
Molecular Formula: C10H15N3S
InChi Key: FFHDQWVSXSKWLF-UHFFFAOYSA-N
SMILES: NCC1=CC(=NC=C1)N1CCSCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results