accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 886851-55-4

OONOOH

CAS RN 886851-55-4

IUPAC Name: 6-(oxan-4-yloxy)pyridine-3-carboxylic acid
Synonyms: 6-(oxan-4-yloxy)pyridine-3-carboxylic acid
Molecular Weight (g/mol): 223.23
Molecular Formula: C11H13NO4
InChi Key: OLOSQIMMFCSJHD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(OC2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results