accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 886851-57-6

IUPAC Name: [4-methyl-2-(pyridin-4-yl)-1,3-thiazol-5-yl]methanol
Synonyms: [4-methyl-2-(pyridin-4-yl)-1,3-thiazol-5-yl]methanol
Molecular Weight (g/mol): 206.26
Molecular Formula: C10H10N2OS
InChi Key: WSBUFMYJALKFHJ-UHFFFAOYSA-N
SMILES: CC1=C(CO)SC(=N1)C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results