accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 887144-94-7

IUPAC Name: 1-(trifluoromethyl)-3H-1λ³,2-benziodaoxol-3-one
Synonyms: 1-(trifluoromethyl)-1λ³,2-benziodaoxol-3-one
Molecular Weight (g/mol): 316.02
Molecular Formula: C8H4F3IO2
InChi Key: XHEOXSQMBWJOKP-UHFFFAOYSA-N
SMILES: FC(F)(F)[I]1OC(=O)C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results