accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOPOOFFFOFFF

CAS RN 88738-78-7

IUPAC Name: methyl 2-[bis(2,2,2-trifluoroethoxy)phosphoryl]acetate
Synonyms: methyl 2-[bis(2,2,2-trifluoroethoxy)phosphoryl]acetate
Molecular Weight (g/mol): 318.11
Molecular Formula: C7H9F6O5P
InChi Key: PVSJXEDBEXYLML-UHFFFAOYSA-N
SMILES: COC(=O)CP(=O)(OCC(F)(F)F)OCC(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results