accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOO(S)(S)FFFFF

CAS RN 88744-04-1

IUPAC Name: (1S,6S)-1-[(9H-fluoren-9-yl)methyl]-2,3,4,5,6-pentafluorocyclohexa-2,4-dien-1-yl hydrogen carbonate
Synonyms: (1S,6S)-1-(9H-fluoren-9-ylmethyl)-2,3,4,5,6-pentafluorocyclohexa-2,4-dien-1-yl hydrogen carbonate
Molecular Weight (g/mol): 408.32
Molecular Formula: C21H13F5O3
InChi Key: YQCQZAIPKWXXJX-NQIIRXRSSA-N
SMILES: OC(=O)O[C@@]1(CC2C3=CC=CC=C3C3=CC=CC=C23)[C@H](F)C(F)=C(F)C(F)=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results