accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 887707-25-7

NIClFFF

CAS RN 887707-25-7

IUPAC Name: 2-chloro-5-iodo-3-(trifluoromethyl)pyridine
Synonyms: 2-chloro-5-iodo-3-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 307.44
Molecular Formula: C6H2ClF3IN
InChi Key: KUASPTUGCBGXAE-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(I)=CN=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results