accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOOHHCl

CAS RN 887922-93-2

IUPAC Name: 3-[(pyrrolidin-1-yl)methyl]benzoic acid hydrochloride
Synonyms: 3-(pyrrolidin-1-ylmethyl)benzoic acid hydrochloride
Molecular Weight (g/mol): 241.72
Molecular Formula: C12H16ClNO2
InChi Key: BORITTZEFSYNFT-UHFFFAOYSA-N
SMILES: Cl.OC(=O)C1=CC(CN2CCCC2)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results