accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3OCH3CH3OSr

CAS RN 88863-33-6

IUPAC Name: strontium(2+) bis(propan-2-olate)
Synonyms: strontium(2+) bis(propan-2-olate)
Molecular Weight (g/mol): 205.8
Molecular Formula: C6H14O2Sr
InChi Key: OHULXNKDWPTSBI-UHFFFAOYSA-N
SMILES: [Sr++].CC(C)[O-].CC(C)[O-]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)