accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2OHHOO

CAS RN 89-57-6

IUPAC Name: 5-amino-2-hydroxybenzoic acid
Synonyms: mesalazine antigen brand of mesalamine farmasa brand of mesalamine monosodium salt, mesalamine lialda allphar brand of mesalamine asacol byk brand of mesalamine 5-asa rowasa
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: KBOPZPXVLCULAV-UHFFFAOYSA-N
SMILES: NC1=CC=C(O)C(=C1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (8)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results