accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 89-60-1

CH3ClNOO

CAS RN 89-60-1

IUPAC Name: 1-chloro-4-methyl-2-nitrobenzene
Synonyms: 1-chloro-4-methyl-2-nitrobenzene
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: NWESJZZPAJGHRZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(Cl)C(=C1)[N+]([O-])=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results