accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH2NOO

CAS RN 89-62-3

IUPAC Name: 4-methyl-2-nitroaniline
Synonyms: 2-nitro-P-toluidine
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: DLURHXYXQYMPLT-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C(=C1)[N+]([O-])=O
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results