accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOH3C

CAS RN 89-71-4

IUPAC Name: methyl 2-methylbenzoate
Synonyms: O-toluic acid, methyl ester
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: WVWZECQNFWFVFW-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1C
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)