accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3(R)CH3H3CO

CAS RN 89-82-7

IUPAC Name: (5R)-5-methyl-2-(propan-2-ylidene)cyclohexan-1-one
Synonyms: (+)-pulegone (+)-(R)-pulegone pulegone (natural) 4(8)-p-menthen-3-one, delta- D-pulegone (R)-pulegone (R)-(+)-pulegone pulegon delta-4(8)-p-menthen-3-one d-pulegone
Molecular Weight (g/mol): 152.24
Molecular Formula: C10H16O
InChi Key: NZGWDASTMWDZIW-MRVPVSSYSA-N
SMILES: C[C@@H]1CCC(=C(C)C)C(=O)C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results