accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3CH3HO

CAS RN 89-83-8

IUPAC Name: 5-methyl-2-(propan-2-yl)phenol
Synonyms: thymol fema number 3066 epa pesticide chemical code 080402 isopropyl-m-cresol thymol preparation thyme camphor m-thymol thymol (natural) thymol (substance) isopropyl cresol
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: MGSRCZKZVOBKFT-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(C)C=C1O
Molecular Weight (g/mol) 
  • (7)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Boiling Point 
  • (1)
  • (3)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (7)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)