accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3NCS

CAS RN 89007-45-4

IUPAC Name: 1-isothiocyanato-4-(propan-2-yl)benzene
Synonyms: 1-isopropyl-4-isothiocyanatobenzene
Molecular Weight (g/mol): 177.27
Molecular Formula: C10H11NS
InChi Key: SNWJJTIJIDXIHL-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(C=C1)N=C=S
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results