accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BFFFFFFFFFFFFFFFFFFFFK

CAS RN 89171-23-3

IUPAC Name: potassium tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
Synonyms: potassium tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
Molecular Weight (g/mol): 718.14
Molecular Formula: C24BF20K
InChi Key: GYBHRIJOPWTIKA-UHFFFAOYSA-N
SMILES: [K+].FC1=C(F)C(F)=C(C(F)=C1F)[B-](C1=C(F)C(F)=C(F)C(F)=C1F)(C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results