CAS RN 892501-99-4
IUPAC Name:
4-[5-(trifluoromethyl)pyridin-2-yl]benzonitrile
Synonyms:
4-[5-(trifluoromethyl)pyridin-2-yl]benzonitrile
Molecular Weight (g/mol):
248.21
Molecular Formula:
C13H7F3N2
InChi Key:
DFPKSXZRVOQLJC-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CN=C(C=C1)C1=CC=C(C=C1)C#N
1
–
1
of
1
results
1
4-[5-(Trifluoromethyl)pyrid-2-yl]benzonitrile, 97%, Thermo Scientific™
CAS: 892501-99-4 Molecular Formula: C13H7F3N2 Molecular Weight (g/mol): 248.21 MDL Number: MFCD09064969 InChI Key: DFPKSXZRVOQLJC-UHFFFAOYSA-N Synonym: 4-5-trifluoromethyl pyridin-2-yl benzonitrile,4-5-trifluoromethyl pyrid-2-yl benzonitrile,4-5-trifluoromethyl-2-pyridinyl benzonitrile,benzonitrile,4-5-trifluoromethyl-2-pyridinyl,4-5-trifluoromethyl-2-pyridyl benzenecarbonitrile,4-5-trifluoromethyl pyridin-2-yl benzenecarbonitrile PubChem CID: 24229535 IUPAC Name: 4-[5-(trifluoromethyl)pyridin-2-yl]benzonitrile SMILES: FC(F)(F)C1=CN=C(C=C1)C1=CC=C(C=C1)C#N