accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 892502-09-9

H2NNN

CAS RN 892502-09-9

IUPAC Name: 3-[(1H-pyrazol-1-yl)methyl]aniline
Synonyms: 3-(pyrazol-1-ylmethyl)aniline
Molecular Weight (g/mol): 173.22
Molecular Formula: C10H11N3
InChi Key: JGTYTUGTENJXBY-UHFFFAOYSA-N
SMILES: NC1=CC=CC(CN2C=CC=N2)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results