accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 892502-27-1

IUPAC Name: 2-(2-methyl-1H-imidazol-1-yl)benzonitrile
Synonyms: 2-(2-methylimidazol-1-yl)benzonitrile
Molecular Weight (g/mol): 183.21
Molecular Formula: C11H9N3
InChi Key: NCQQCFZREDABTC-UHFFFAOYSA-N
SMILES: CC1=NC=CN1C1=CC=CC=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results