accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 89268-62-2

CAS RN 89268-62-2

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results