accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 893-33-4

IUPAC Name: 4,4,4-trifluoro-1-(naphthalen-2-yl)butane-1,3-dione
Synonyms: 4,4,4-trifluoro-1-(naphthalen-2-yl)butane-1,3-dione
Molecular Weight (g/mol): 266.22
Molecular Formula: C14H9F3O2
InChi Key: WVVLURYIQCXPIV-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)CC(=O)C1=CC=C2C=CC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results