accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 893440-50-1

IUPAC Name: 2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-amine
Synonyms: 2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-amine
Molecular Weight (g/mol): 250.11
Molecular Formula: C12H19BN2O3
InChi Key: KYYKGOURQXPERA-UHFFFAOYSA-N
SMILES: COC1=C(N)C=C(C=N1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results