accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 89359-54-6

IUPAC Name: 9-bromonon-1-ene
Synonyms: 9-bromonon-1-ene
Molecular Weight (g/mol): 205.14
Molecular Formula: C9H17Br
InChi Key: RQXPBVHYVAOUBY-UHFFFAOYSA-N
SMILES: BrCCCCCCCC=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results