CAS RN 89365-50-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Salmeterol 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Salmeterol, TRC
CAS: 89365-50-4 Molecular Formula: C25 H37 N O4 Molecular Weight (g/mol): 415.57 Synonym: 1,3-Benzenedimethanol, 4-hydroxy-α1-[[[6-(4-phenylbutoxy)hexyl]amino]methyl]-,1,3-Benzenedimethanol, 4-hydroxy-α1-[[[6-(4-phenylbutoxy)hexyl]amino]methyl]-, (±)-,GR 33343X,Salmeterol IUPAC Name: 2-(hydroxymethyl)-4-[1-hydroxy-2-[6-(4-phenylbutoxy)hexylamino]ethyl]phenol SMILES: OCc1cc(ccc1O)C(O)CNCCCCCCOCCCCc2ccccc2