accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 893741-66-7

IUPAC Name: methyl({5-[(morpholin-4-yl)methyl]furan-2-yl}methyl)amine
Synonyms: methyl({[5-(morpholin-4-ylmethyl)furan-2-yl]methyl})amine
Molecular Weight (g/mol): 210.28
Molecular Formula: C11H18N2O2
InChi Key: RVGSMPQAXGUMNM-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(CN2CCOCC2)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results