accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NHSNO

CAS RN 893742-71-7

IUPAC Name: methyl({5-[(morpholin-4-yl)methyl]thiophen-2-yl}methyl)amine
Synonyms: methyl({[5-(morpholin-4-ylmethyl)thiophen-2-yl]methyl})amine
Molecular Weight (g/mol): 226.34
Molecular Formula: C11H18N2OS
InChi Key: LJPRPFHZPNCEIQ-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(CN2CCOCC2)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results