CAS RN 893748-66-8
IUPAC Name:
1-{5-[(morpholin-4-yl)methyl]-1,2-oxazol-3-yl}methanamine
Synonyms:
1-[5-(morpholin-4-ylmethyl)-1,2-oxazol-3-yl]methanamine
Molecular Weight (g/mol):
197.24
Molecular Formula:
C9H15N3O2
InChi Key:
JJKIQRMZVVZXSK-UHFFFAOYSA-N
SMILES:
NCC1=NOC(CN2CCOCC2)=C1
1
–
1
of
1
results
1
[5-(Morpholinomethyl)isoxazol-3-yl]methylamine, 97%, Thermo Scientific™
CAS: 893748-66-8 Molecular Formula: C9H15N3O2 Molecular Weight (g/mol): 197.24 MDL Number: MFCD06804185 InChI Key: JJKIQRMZVVZXSK-UHFFFAOYSA-N Synonym: 5-morpholinomethyl isoxazol-3-yl methylamine,3-isoxazolemethanamine, 5-4-morpholinylmethyl,5-morpholin-4-ylmethyl-1,2-oxazol-3-yl methanamine,1-5-morpholin-4-ylmethyl-1,2-oxazol-3-yl methanamine PubChem CID: 20099025 IUPAC Name: [5-(morpholin-4-ylmethyl)-1,2-oxazol-3-yl]methanamine SMILES: C1COCCN1CC2=CC(=NO2)CN