CAS RN 894351-84-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(1E)-N-Methyl-2-[1-(phenylmethyl)-1H-indol-5-yl]ethenesulfonamide, TRC
CAS: 894351-84-9 Molecular Formula: C18 H18 N2 O2 S Molecular Weight (g/mol): 326.41 IUPAC Name: (E)-2-(1-benzylindol-5-yl)-N-methylethenesulfonamide SMILES: CNS(=O)(=O)\C=C\c1ccc2c(ccn2Cc3ccccc3)c1