accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 894406-63-4

IUPAC Name: 2-(trifluoromethyl)-[1,3]oxazolo[4,5-b]pyridine
Synonyms: 2-(trifluoromethyl)-[1,3]oxazolo[4,5-b]pyridine
Molecular Weight (g/mol): 188.11
Molecular Formula: C7H3F3N2O
InChi Key: MJILPMZYOJELAX-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=NC2=NC=CC=C2O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results