accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOBOHHO

CAS RN 89466-08-0

IUPAC Name: (2-hydroxyphenyl)boronic acid
Synonyms: 2-hydroxyphenylboronic acid
Molecular Weight (g/mol): 137.93
Molecular Formula: C6H7BO3
InChi Key: YDMRDHQUQIVWBE-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC=C1O
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)