accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 89570-82-1

IUPAC Name: 3-chloro-2-hydrazinyl-5-(trifluoromethyl)pyridine
Synonyms: 3-chloro-2-hydrazinyl-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 211.57
Molecular Formula: C6H5ClF3N3
InChi Key: KHFKSHYCVQZAQP-UHFFFAOYSA-N
SMILES: NNC1=NC=C(C=C1Cl)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results