CAS RN 89581-58-8
IUPAC Name:
2-chloro-6-methylpyrimidine-4-carboxylic acid
Synonyms:
2-chloro-6-methylpyrimidine-4-carboxylic acid
Molecular Weight (g/mol):
172.57
Molecular Formula:
C6H5ClN2O2
InChi Key:
FBXGLOGQWMGIER-UHFFFAOYSA-N
SMILES:
CC1=NC(Cl)=NC(=C1)C(O)=O
1
–
1
of
1
results
1
2-chloro-6-methylpyrimidine-4-carboxylic acid, 95%, Thermo Scientific Chemicals
CAS: 89581-58-8 Molecular Formula: C6H5ClN2O2 Molecular Weight (g/mol): 172.57 MDL Number: MFCD00833002 InChI Key: FBXGLOGQWMGIER-UHFFFAOYSA-N Synonym: 2-chloro-6-methyl-pyrimidine-4-carboxylic acid,2-chloro-6-methyl-4-pyrimidinecarboxylic acid,2-chloro-4-methylpyrimidine-6-carboxylic acid,4-pyrimidinecarboxylic acid, 2-chloro-6-methyl PubChem CID: 10535213 IUPAC Name: 2-chloro-6-methylpyrimidine-4-carboxylic acid SMILES: CC1=NC(=NC(=C1)C(=O)O)Cl