accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 89598-96-9

IUPAC Name: (3-bromophenyl)boronic acid
Synonyms: 3-bromophenylboronic acid
Molecular Weight (g/mol): 200.83
Molecular Formula: C6H6BBrO2
InChi Key: AFSSVCNPDKKSRR-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC(Br)=C1
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results