accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BBrOHHOOH

CAS RN 89598-97-0

IUPAC Name: (5-bromo-2-hydroxyphenyl)boronic acid
Synonyms: 5-bromo-2-hydroxyphenylboronic acid
Molecular Weight (g/mol): 216.83
Molecular Formula: C6H6BBrO3
InChi Key: XSXTWLOHAGPBNO-UHFFFAOYSA-N
SMILES: OB(O)C1=C(O)C=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results