accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 89694-44-0

BOH3CBrHOOH

CAS RN 89694-44-0

IUPAC Name: (2-bromo-5-methoxyphenyl)boronic acid
Synonyms: 2-bromo-5-methoxyphenylboronic acid
Molecular Weight (g/mol): 230.85
Molecular Formula: C7H8BBrO3
InChi Key: BJQKEDXKQVNQPR-UHFFFAOYSA-N
SMILES: COC1=CC(B(O)O)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results