accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 89694-48-4

BClOCH3HOOH

CAS RN 89694-48-4

IUPAC Name: (5-chloro-2-methoxyphenyl)boronic acid
Synonyms: 5-chloro-2-methoxyphenylboronic acid
Molecular Weight (g/mol): 186.4
Molecular Formula: C7H8BClO3
InChi Key: FMBVAOHFMSQDGT-UHFFFAOYSA-N
SMILES: COC1=C(C=C(Cl)C=C1)B(O)O
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)