accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NClClFFF

CAS RN 89719-92-6

IUPAC Name: 2,5-dichloro-4-(trifluoromethyl)pyridine
Synonyms: 2,5-dichloro-4-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 215.98
Molecular Formula: C6H2Cl2F3N
InChi Key: GFTRYJQAASFSIB-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=NC=C1Cl
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results