accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FOFFF

CAS RN 89763-93-9

IUPAC Name: 2-fluoro-4-(trifluoromethyl)benzaldehyde
Synonyms: 2-fluoro-4-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 192.11
Molecular Formula: C8H4F4O
InChi Key: KFEHNXLFIGPWNB-UHFFFAOYSA-N
SMILES: FC1=CC(=CC=C1C=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (8)
  • (3)
Percent Purity 
  • (9)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
Boiling Point 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (3)
  • (6)
  • (8)
  • (3)
  • (9)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)