accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3(S)NNN(S)N(R)OSOOHOO

CAS RN 89786-04-9

IUPAC Name: (2S,3S,5R)-3-methyl-4,4,7-trioxo-3-[(1H-1,2,3-triazol-1-yl)methyl]-4λ⁶-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Synonyms: tazobactam
Molecular Weight (g/mol): 300.29
Molecular Formula: C10H12N4O5S
InChi Key: LPQZKKCYTLCDGQ-WEDXCCLWSA-N
SMILES: C[C@]1(CN2C=CN=N2)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(O)=O
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)