accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNOOH3CCl

CAS RN 89793-11-3

IUPAC Name: methyl 2-chloro-6-methylpyrimidine-4-carboxylate
Synonyms: methyl 2-chloro-6-methylpyrimidine-4-carboxylate
Molecular Weight (g/mol): 186.6
Molecular Formula: C7H7ClN2O2
InChi Key: ZNPORLVPCMFKOR-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(C)=NC(Cl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results